Product Name
3,5-Dimethylpyrazole-1-carboxamide
Product Number
D182206
Product Brand
ALDRICH
CAS Number
934-48-5
Molecular Weight
139.16
Test | Specification |
---|---|
APPEARANCE | WHITE CRYSTALS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON TYPICALLY 47.9% - 54.9% |
NITROGEN TYPICALLY 28.4% - 32.0% | |
GAS LIQUID | TYPICALLY >98% |
APPROVED MARCH 27, 2002 JSB |