Product Name
6-Aminoindazole 98%
Product Number
A59565
Product Brand
ALDRICH
CAS Number
6967-12-0
Molecular Weight
133.15
Test | Specification |
---|---|
APPEARANCE | WHITE TO BROWN POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 61.5%-64.7% |
NITROGEN 30.7%-32.3% | |
HIGH PRESSURE LIQUID | 97.5% (MINIMUM) |
APPROVED OCTOBER 28, 2005 RJM |