Product Name
(3-Hydroxy-1-propenyl)sulfur pentafluoride technical grade
Product Number
697648
Product Brand
ALDRICH
CAS Number
155990-90-2
Molecular Weight
184.13
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO TAN LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 88.0% (MINIMUM) |
APPROVED MAY 11, 2008 DJP |