Product Name
3-Methyl-4-nitrobenzhydrazide 97%
Product Number
680370
Product Brand
ALDRICH
CAS Number
72198-83-5
Molecular Weight
195.18
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN POWDER, CRYSTALS OR CRYSTALLINE |
POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 47.5% - 51.0% |
NITROGEN 20.7% - 22.3% | |
AND/OR | |
TITRATION | 96.5% - 103.5% (WITH HCLO4) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED FEBRUARY 8, 2007 BTR |