Product Name
5-Chlorothiophene-2-carboxylic acid hydrazide 97%
Product Number
678619
Product Brand
ALDRICH
CAS Number
351983-31-8
Molecular Weight
176.62
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN SOLID, POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5-103.5% (WITH HCLO4) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED MAY 17, 2007 DJP |