Product Name
4-(4-Methylphenyl)benzaldehyde 97%
Product Number
664782
Product Brand
ALDRICH
CAS Number
36393-42-7
Molecular Weight
196.24
Test | Specification |
---|---|
APPEARANCE | WHITE TO YELLOW POWDER, CRYSTALS OR |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5% - 103.5% (WITH HYDROXYLAMINE) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED APRIL 4, 2006 BTR |