Product Name
5-(4-Chlorophenyl)oxazole-2-propionic acid 97%
Product Number
658553
Product Brand
ALDRICH
CAS Number
23464-95-1
Molecular Weight
251.67
Test | Specification |
---|---|
APPEARANCE | WHITE TO YELLOW TO TAN POWDER, CRYSTALS |
OR CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 55.3% - 59.3% |
NITROGEN 5.3% - 5.8% | |
AND/OR | |
TITRATION | 96.5% - 103.5% (WITH AGNO3 AFTER OXYGEN |
COMBUSTION) | |
HIGH PRESSURE LIQUID | 96.5% (MINIMUM) |
APPROVED JUNE 29, 2005 |