Product Name
2-Mercapto-6-methylpyridine 97%
Product Number
642096
Product Brand
ALDRICH
CAS Number
18368-57-5
Molecular Weight
125.19
Test | Specification |
---|---|
Appearance (Color) | White to Yellow |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Proton NMR spectra | Conforms to Structure |
Carbon | 55.6 - 59.6 % |
Nitrogen | 10.8 - 11.6 % |
Purity (GC) | ≥96.5 % |