Product Name
4-(2-Pyridyl)benzaldehyde 99%
Product Number
572411
Product Brand
ALDRICH
CAS Number
127406-56-8
Molecular Weight
183.21
Test | Specification |
---|---|
Appearance (Color) | White |
Appearance (Form) | Crystal to Powder |
Infrared spectrum | Conforms to Structure |
Carbon | 77.4 - 79.9 % |
Nitrogen | 7.3 - 7.9 % |
Purity (HPLC) | ≥98.5 % |