Product Name
5-Phenylthiophene-2-carboxaldehyde 98%
Product Number
565938
Product Brand
ALDRICH
CAS Number
19163-21-4
Molecular Weight
188.25
Test | Specification |
---|---|
Appearance (Color) | Yellow to Orange |
Appearance (Form) | Conforms to Requirements |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Titration by Hydroxylamine | 97.5 - 102.5 % |
Purity (HPLC) | ≥97.5 % |