Product Name
4-Bromo-2-chlorotoluene 98%
Product Number
528889
Product Brand
ALDRICH
CAS Number
89794-02-5
Molecular Weight
205.48
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 97.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |