Product Name
N-Methylpentylamine 98%
Product Number
496138
Product Brand
ALDRICH
CAS Number
25419-06-1
Molecular Weight
101.19
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
TITRATION | 97.5%-102.5% (WITH HCL) |
GAS LIQUID | 97.5% (MINIMUM) |
APPROVED OCTOBER 30, 2008 RJM |