Product Name
4-(Difluoromethoxy)benzoic acid 98%
Product Number
470104
Product Brand
ALDRICH
CAS Number
4837-20-1
Molecular Weight
188.13
Test | Specification |
---|---|
Appearance (Color) | White |
Appearance (Form) | Conforms to Requirements |
Powder, Crystals or Flakes | |
Infrared spectrum | Conforms to Structure |
Purity (Titration by NaOH) | 97.5 - 102.5 % |
Purity (TLC) | ≥98 % |
Solubility (Turbidity) | Clear |
50mg/ml, Methanol | |
Solubility (Color) | Colorless |