Product Name
1-(2-Fluorophenyl)piperazine 97%
Product Number
444804
Product Brand
ALDRICH
CAS Number
1011-15-0
Molecular Weight
180.22
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid or Solid
TITRATION (NT) HCLO4 0.1M96.5 - 103.5 %
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE