Product Name
4,4′-Bis[(4-chlorophenyl)sulfonyl]-1,1′-biphenyl 98%
Product Number
440949
Product Brand
ALDRICH
CAS Number
22287-56-5
Molecular Weight
503.42
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5-102.5% (WITH AGNO3 AFTER O2 COMBUSTION) |
APPROVED DECEMBER 5, 2007 RJM |