Product Name
4-Isothiocyanatophenyl 4-pentylbicyclo[2.2.2]octane-1-carboxylate liquid crystal (nematic), 99%
Product Number
370053
Product Brand
ALDRICH
CAS Number
121235-90-3
Molecular Weight
357.51
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER, CRYSTALS AND/OR CHUNKS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 69.5%-71.6% |
NITROGEN 3.7%- 4.1% | |
HIGH PRESSURE LIQUID | 98.5% (MINIMUM) |
APPROVED FEBRUARY 9, 2009 RJM |