Product Name
Zinc carbonate hydroxide hydrate 97%
Product Number
367206
Product Brand
ALDRICH
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER |
X-RAY DIFFRACTION | CONFORMS TO STANDARD PATTERN. |
OR | |
ICP ASSAY | CONFIRMS ZINC COMPONENT. |
TITRATION | TYPICALLY 55% - 58% ZN (COMPLEXOMETRIC) |
APPROVED MAY 16, 1997 JSB | |
REVISED, JUNE 16, 2000 DJS |