Product Name
Butyltin hydroxide oxide hydrate 97%
Product Number
344931
Product Brand
ALDRICH
CAS Number
336879-56-2
Molecular Weight
208.83 (anhydrous basis)
Test | Specification |
---|---|
Appearance (Color) | White |
Appearance (Form) | Conforms to Requirement |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Carbon (dry basis) | 22.2 - 23.8 % |
Water (by Karl Fischer) | ≥4.1 % |