Product Name
1,5-Dicyclohexylimidazole 97%
Product Number
316547
Product Brand
ALDRICH
CAS Number
80964-44-9
Molecular Weight
232.36
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER OR CRYSTALS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5%-103.5% (WITH HCLO4) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED SEPTEMBER 17, 2008 RJM |