Product Name
4-Sulfobenzoic acid potassium salt 95%
Product Number
310638
Product Brand
ALDRICH
CAS Number
5399-63-3
Molecular Weight
240.27
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder or Crystals or Flakes |
TITRATION (T) NAOH 0.1M | 94.0 - 106.0 % |
PURITY (TLC AREA %) | ≥ 94.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |