Product Name
Ethyl 3,5-dinitrobenzoate 99%
Product Number
301337
Product Brand
ALDRICH
CAS Number
618-71-3
Molecular Weight
240.17
Test | Specification |
---|---|
Appearance (Color) | White to Yellow |
Appearance (Form) | Conforms to Requirements |
Powder, Crystals or Fibers | |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥98.5 % |