Product Name
Trichloromethyl isocyanate
Product Number
250767
Product Brand
ALDRICH
CAS Number
30121-98-3
Molecular Weight
160.39
Test | Specification |
---|---|
Appearance (Color) | Colorless |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | % |
Typically 97% - 103% After Oxygen Combustion | |
Purity (GC) | % |
Typically >85% |