Skip to Content

V001098

2,6-Dichloroindophenol sodium salt hydrate

98%

Synonym(s):

2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt, 2,6-Dichlorobenzenone-indophenol sodium salt, 2,6-Dichlorophenolindophenol sodium salt hydrate, DCIP, DPIP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H6Cl2NNaO2 · xH2O
CAS Number:
Molecular Weight:
290.08 (anhydrous basis)
EC Number:
210-640-4
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3641229
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

98%

SMILES string

O.[Na+].[O-]c1ccc(cc1)\N=C2/C=C(Cl)C(=O)C(Cl)=C2

InChI

1S/C12H7Cl2NO2.Na.H2O/c13-10-5-8(6-11(14)12(10)17)15-7-1-3-9(16)4-2-7;;/h1-6,16H;;1H2/q;+1;/p-1

InChI key

XHSOLXWCGCVQHE-UHFFFAOYSA-M



Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library