Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H98772608

Targetmol Chemicals Inc.

Verilopam

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
326.44
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

4-[2-(7,8-dimethoxy-1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)ethyl]aniline

Inchi Code

1S/C20H26N2O2/c1-23-19-13-16-8-11-22(12-9-17(16)14-20(19)24-2)10-7-15-3-5-18(21)6-4-15/h3-6,13-14H,7-12,21H2,1-2H3

InChI key

BTCHMHMCMKZOQS-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

97.91%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Verilopam

MSDS

https://www.targetmol.com/compound/Verilopam


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service