Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H98048BA7

Targetmol Chemicals Inc.

ML-178

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
441.93
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

2,6-dibromo-3-(2-(2,4-dichlorophenoxy)ethoxy)pyridine

Inchi Code

1S/C13H9Br2Cl2NO2/c14-12-4-3-11(13(15)18-12)20-6-5-19-10-2-1-8(16)7-9(10)17/h1-4,7H,5-6H2

InChI key

KMGUBUXDNOMSRM-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

95.92%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/ML-178

MSDS

https://www.targetmol.com/compound/ML-178


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service