Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H97F3147B

Targetmol Chemicals Inc.

SDZ WAG 994

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
363.42

Iupac Name

(2R,3R,4R,5R)-5-(6-(cyclohexylamino)-9H-purin-9-yl)-2-(hydroxymethyl)-4-methoxytetrahydrofuran-3-ol

Inchi Code

1S/C17H25N5O4/c1-25-14-13(24)11(7-23)26-17(14)22-9-20-12-15(18-8-19-16(12)22)21-10-5-3-2-4-6-10/h8-11,13-14,17,23-24H,2-7H2,1H3,(H,18,19,21)/t11-,13-,14-,17-/m1/s1

InChI key

JAKAFSGZUXCHLF-LSCFUAHRSA-N

storage temp.

-80/-20

Purity

99.90%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/SDZ WAG 994

MSDS

https://www.targetmol.com/compound/SDZ WAG 994


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service