Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H97BAF23D

Targetmol Chemicals Inc.

R715 TFA(185052-09-9 free base)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$152.00
5 MG
$326.00
10 MG
$490.00
25 MG
$783.00
50 MG
$1,090.00
100 MG
$1,480.00

Vendor SKU: TP1933-1MG

$152.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 MG
$152.00
5 MG
$326.00
10 MG
$490.00
25 MG
$783.00
50 MG
$1,090.00
100 MG
$1,480.00

About This Item

Molecular Weight:
1254.37

Vendor SKU: TP1933-1MG

$152.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

((R)-2-((S)-2-((S)-2-(2-((S)-1-(((S)-2-((S)-2-acetamido-6-aminohexanamido)-5-((diaminomethylene)amino)pentanoyl)-L-prolyl)pyrrolidine-2-carboxamido)acetamido)-3-phenylpropanamido)-3-hydroxypropanamido)-3-(naphthalen-2-yl)propanoyl)-L-isoleucine--2,2,2-trifluoroacetic acid (1/1)

Inchi Code

1S/C57H81N13O12.C2HF3O2/c1-4-34(2)48(56(81)82)68-51(76)43(31-37-23-24-38-17-8-9-18-39(38)29-37)66-52(77)44(33-71)67-50(75)42(30-36-15-6-5-7-16-36)64-47(73)32-62-53(78)45-21-13-27-69(45)55(80)46-22-14-28-70(46)54(79)41(20-12-26-61-57(59)60)65-49(74)40(63-35(3)72)19-10-11-25-58;3-2(4,5)1(6)7/h5-9,15-18,23-24,29,34,40-46,48,71H,4,10-14,19-22,25-28,30-33,58H2,1-3H3,(H,62,78)(H,63,72)(H,64,73)(H,65,74)(H,66,77)(H,67,75)(H,68,76)(H,81,82)(H4,59,60,61);(H,6,7)/t34-,40-,41-,42-,43+,44-,45-,46-,48-;/m0./s1

Inchi Key

ZFUJDXXJCMFOSK-WBUMRSSGSA-N

Storage temp.

-80/-20

Purity

100%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/R715 TFA(185052-09-9 free base)

MSDS

https://www.targetmol.com/compound/R715 TFA(185052-09-9 free base)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service