Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H97BAF171

Targetmol Chemicals Inc.

H-Gly-Gly-Pro-OH

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$34.00
25 MG
$54.00
50 MG
$77.00
100 MG
$113.00
200 MG
$166.00

Vendor SKU: TP1156-10MG

$34.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
10 MG
$34.00
25 MG
$54.00
50 MG
$77.00
100 MG
$113.00
200 MG
$166.00

About This Item

CAS Number:
Molecular Weight:
229.24

Vendor SKU: TP1156-10MG

$34.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

(2S)-1-{[(aminoacetyl)amino]acetyl}-2-pyrrolidinecarboxylic acid

Inchi Code

1S/C9H15N3O4/c10-4-7(13)11-5-8(14)12-3-1-2-6(12)9(15)16/h6H,1-5,10H2,(H,11,13)(H,15,16)/t6-/m0/s1

InChI key

BUEFQXUHTUZXHR-LURJTMIESA-N

storage temp.

-80/-20

Purity

97.44%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/H-Gly-Gly-Pro-OH

MSDS

https://www.targetmol.com/compound/H-Gly-Gly-Pro-OH


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service