Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H97BAF05D

Targetmol Chemicals Inc.

NAcM-OPT HCl(2089293-61-6 free base)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
470.87

Iupac Name

1-benzyl-1-(1-butylpiperidin-4-yl)-3-(3,4-dichlorophenyl)urea hydrochloride

Inchi Code

1S/C23H29Cl2N3O.ClH/c1-2-3-13-27-14-11-20(12-15-27)28(17-18-7-5-4-6-8-18)23(29)26-19-9-10-21(24)22(25)16-19;/h4-10,16,20H,2-3,11-15,17H2,1H3,(H,26,29);1H

InChI key

DDYSTAUKIBWFNE-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

99.43%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/NAcM-OPT HCl(2089293-61-6 free base)

MSDS

https://www.targetmol.com/compound/NAcM-OPT HCl(2089293-61-6 free base)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service