TA9H94532C1E
Targetmol Chemicals Inc.
1,4-Chrysenequinone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
258.28
Iupac Name
1,4-chrysenedione
Inchi Code
1S/C18H10O2/c19-16-9-10-17(20)18-14-6-5-11-3-1-2-4-12(11)13(14)7-8-15(16)18/h1-10H
InChI key
UORKIKBNUWJNJF-UHFFFAOYSA-N
storage temp.
-80/-20
Purity
99.65%
Country of Origin
CN
Shipping Temperature
4
Physical Form
solid
Product Link
https://www.targetmol.com/compound/1,4-Chrysenequinone
MSDS
https://www.targetmol.com/compound/1,4-Chrysenequinone
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service