Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H93ED6CC1

Targetmol Chemicals Inc.

Eltoprazine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
293.19

Iupac Name

1-(2,3-dihydrobenzo[b][1,4]dioxin-5-yl)piperazine dihydrochloride

Inchi Code

1S/C12H16N2O2.2ClH/c1-2-10(14-6-4-13-5-7-14)12-11(3-1)15-8-9-16-12;;/h1-3,13H,4-9H2;2*1H

InChI key

UNRJDVNCYJTPOQ-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

99.94%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Eltoprazine dihydrochloride

MSDS

https://www.targetmol.com/compound/Eltoprazine dihydrochloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service