Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H93CFBF7A

Targetmol Chemicals Inc.

9''-Methyl salvianolate B

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$64.00
5 MG
$147.00
10 MG
$218.00
25 MG
$369.00
50 MG
$543.00
100 MG
$776.00

Vendor SKU: T4961-1MG

$64.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 MG
$64.00
5 MG
$147.00
10 MG
$218.00
25 MG
$369.00
50 MG
$543.00
100 MG
$776.00

About This Item

CAS Number:
Molecular Weight:
732.65

Vendor SKU: T4961-1MG

$64.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

(2R)-2-({(2E)-3-[(2S,3S)-3-({[(1R)-1-(3,4-dihydroxybenzyl)-2-methoxy-2-oxoethyl]oxy}carbonyl)-2-(3,4-dihydroxyphenyl)-7-hydroxy-2,3-dihydro-1-benzofuran-4-yl]-2-propenoyl}oxy)-3-(3,4-dihydroxyphenyl)propanoic acid

Inchi Code

1S/C37H32O16/c1-50-36(48)29(15-18-3-8-22(39)26(43)13-18)52-37(49)32-31-19(4-10-24(41)34(31)53-33(32)20-5-9-23(40)27(44)16-20)6-11-30(45)51-28(35(46)47)14-17-2-7-21(38)25(42)12-17/h2-13,16,28-29,32-33,38-44H,14-15H2,1H3,(H,46,47)/b11-6+/t28-,29-,32+,33-/m1/s1

Inchi Key

REHAMWBRZKUHPN-NQBFDTSGSA-N

Storage temp.

-80/-20

Purity

97.25%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/9''-Methyl salvianolate B

MSDS

https://www.targetmol.com/compound/9''-Methyl salvianolate B


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service