Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SY3H996B9230

Synthonix Corporation

7-chloro-1H,2H,3H-pyrido[3,4-b][1,4]oxazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
170.6
MDL number:

Iupac Name

7-chloro-2,3-dihydro-1H-pyrido[3,4-b][1,4]oxazine

Inchi Code

1S/C7H7ClN2O/c8-7-3-5-6(4-10-7)11-2-1-9-5/h3-4,9H,1-2H2

InChI key

YRTCKHNUFOCIEP-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/77853.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service