Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SY3H94E1CB20

Synthonix Corporation

n-Decylzinc bromide, 0.50 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$218.50
50 ML
$304.75
100 ML
$519.80

Vendor SKU: D73284-25ML

$218.50


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
25 ML
$218.50
50 ML
$304.75
100 ML
$519.80

About This Item

CAS Number:
Molecular Weight:
286.57
MDL number:

Vendor SKU: D73284-25ML

$218.50


Ships in 10 days - from Aldrich Partner.

Iupac Name

bromo(decyl)zinc

Inchi Code

1S/C10H21.BrH.Zn/c1-3-5-7-9-10-8-6-4-2;;/h1,3-10H2,2H3;1H;/q;;+1/p-1

InChI key

YRORUGDGABRPQJ-UHFFFAOYSA-M

storage temp.

Refrigerator

Purity

0.50

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solution

Product Link

https://synthonix.com/73284.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service