SY3H3D681CE9
Synthonix Corporation
1,1,1,3,3-Pentamethyl-3-phenyldisiloxane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1,1,1,3,3-pentamethyl-3-phenyldisiloxane
Inchi Code
1S/C11H20OSi2/c1-13(2,3)12-14(4,5)11-9-7-6-8-10-11/h6-10H,1-5H3
InChI key
AMNQQNTZDFYVGH-UHFFFAOYSA-N
storage temp.
Refrigerator
Purity
95%
Country of Origin
US
Shipping Temperature
Room Temperature
Physical Form
Colorless to light-yellow liquid
Product Link
https://synthonix.com/55095.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://synthonix.com/55095.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service