Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SY3H3D6817E4

Synthonix Corporation

4-(Piperazin-1-yl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
178.23
MDL number:

Iupac Name

4-(1-piperazinyl)phenol

Inchi Code

1S/C10H14N2O/c13-10-3-1-9(2-4-10)12-7-5-11-6-8-12/h1-4,11,13H,5-8H2

InChI key

GPEOAEVZTOQXLG-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Crystal - Powder / Colour: Very pale yellow - Greyish yellow red

Product Link

https://synthonix.com/39614.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service