SY3H3D680657
Synthonix Corporation
Methyl quinoxaline-5-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 5-quinoxalinecarboxylate
Inchi Code
1S/C10H8N2O2/c1-14-10(13)7-3-2-4-8-9(7)12-6-5-11-8/h2-6H,1H3
InChI key
LCDDKBKKXKEGPK-UHFFFAOYSA-N
storage temp.
Refrigerator
Purity
98%
Country of Origin
US
Shipping Temperature
Room Temperature
Physical Form
Light-green to Pale-yellow to Yellow Solid
Product Link
https://synthonix.com/53660.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://synthonix.com/53660.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service