SY3H3D679AF5
Synthonix Corporation
tert-Butyl ((2S,3S)-4-chloro-3-hydroxy-1-phenylbutan-2-yl)carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl (1S,2S)-1-benzyl-3-chloro-2-hydroxypropylcarbamate
Inchi Code
1S/C15H22ClNO3/c1-15(2,3)20-14(19)17-12(13(18)10-16)9-11-7-5-4-6-8-11/h4-8,12-13,18H,9-10H2,1-3H3,(H,17,19)/t12-,13+/m0/s1
InChI key
GFGQSTIUFXHAJS-QWHCGFSZSA-N
storage temp.
Refrigerator
Purity
95%
Country of Origin
US
Shipping Temperature
Room Temperature
Physical Form
White to off-white powder or crystals
Product Link
https://synthonix.com/58440.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://synthonix.com/58440.html
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service