Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SY3H3249F400

Synthonix Corporation

Ethynylzinc chloride, 0.25M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$276.00
50 ML
$460.00

Vendor SKU: E7775-25ML

$276.00


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
25 ML
$276.00
50 ML
$460.00

About This Item

CAS Number:
Molecular Weight:
125.87
MDL number:

Vendor SKU: E7775-25ML

$276.00


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

ethynylzinc(II) chloride

Inchi Code

1S/C2H.ClH.Zn/c1-2;;/h1H;1H;/q;;+1/p-1

InChI key

OICGZWCIGLUUAM-UHFFFAOYSA-M

storage temp.

Refrigerator

Purity

0.25

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solution

Product Link

https://synthonix.com/7775.html

Signal Word

Danger

Precautionary statements

MSDS

https://synthonix.com/7775.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service