Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SY3428348437

Synthonix Corporation

Methyl-6-aminopyrimidine-4-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
153.14
MDL number:

Iupac Name

methyl 6-amino-4-pyrimidinecarboxylate

Inchi Code

1S/C6H7N3O2/c1-11-6(10)4-2-5(7)9-3-8-4/h2-3H,1H3,(H2,7,8,9)

InChI key

URSUGGQRGURMAM-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

https://synthonix.com/12144.html

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service