Skip to Content
MilliporeSigma

E4628

Ethyl (ethoxymethylene)cyanoacetate

Synonym(s):

Ethyl 2-cyano-3-ethoxyacrylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

EC Number:
202-299-5
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C



Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

266.0 °F - closed cup

flash_point_c

130 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library