Skip to Content

D8635

Diethyl allylmalonate

Synonym(s):

Allylmalonic acid diethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH2=CHCH2CH(COOC2H5)2
CAS Number:
Molecular Weight:
200.23
EC Number:
218-072-9
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
1709466
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


refractive index

n20/D 1.431 (lit.)

bp

222-223 °C (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(CC=C)C(=O)OCC

InChI

1S/C10H16O4/c1-4-7-8(9(11)13-5-2)10(12)14-6-3/h4,8H,1,5-7H2,2-3H3

InChI key

GDWAYKGILJJNBB-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library