Skip to Content
MilliporeSigma

D3773

Veratric acid

Synonym(s):

3,4-Dimethoxybenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3O)2C6H3CO2H
CAS Number:
Molecular Weight:
182.17
EC Number:
202-215-7
UNSPSC Code:
12352209
MDL number:
Beilstein/REAXYS Number:
518285
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

179-182 °C (lit.)

SMILES string

COc1ccc(cc1OC)C(O)=O

InChI

1S/C9H10O4/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5H,1-2H3,(H,10,11)

InChI key

DAUAQNGYDSHRET-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library