Skip to Content
MilliporeSigma

C6663

Cellulose

fibers, long

Synonym(s):

Cellulose powder, Cotton linters

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
EC Number:
232-674-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

fibers

particle size

, long

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Application

Cellulose is a polysaccharide composed of long chains of β(1,4) linked D-glucose units. Cellulose is used in a wide range of applications that require a structural back-bone. It is used in the preparation of chromatographic structural media, in gels for processes such as electrophoresis and ionic separations and as a substrate to identify and study cellulosic systems.
High purity cellulose powders for partition chromatography.


Still not finding the right product?

Explore all of our products under Cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library