Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H12ClNO3
CAS Number:
Molecular Weight:
193.63
UNSPSC Code:
12352200
PubChem Substance ID:
EC Number:
223-827-0
MDL number:
assay
≥99.0% (TLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
CC(C)C(NC(=O)CCl)C(O)=O
InChI
1S/C7H12ClNO3/c1-4(2)6(7(11)12)9-5(10)3-8/h4,6H,3H2,1-2H3,(H,9,10)(H,11,12)
InChI key
LJRISAYPKJORFZ-UHFFFAOYSA-N
Biochem/physiol Actions
N-Chloroacetyl-DL-valine is a racemic mixture of D and L enantiomers of N-chloroacetyl-valine. N-chloroacetyl-L-valine may be used to identify, differentiate and characterize unique N-acylamino acid racemase(s).
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
