Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PFM250

Sigma-Aldrich

GenElute Five-Minute Plasmid Miniprep Kit

sufficient for 250 purifications

Synonym(s):

GenElute Plasmid Miniprep Kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

usage

sufficient for 250 purifications

Looking for similar products? Visit Product Comparison Guide

General description

An overnight recombinant E. coli culture in LB (Luria-Bertani) broth is treated briefly (1-2 minutes) with a lysis reagent for rapid cell lysis and RNA degradation, without prior removal of culture medium. A binding solution is then mixed with the lysate, and DNA is captured on a silica-based binding column. Impurities are removed by a wash solution and bound plasmid DNA is then eluted in Tris buffer or water, ready for immediate use in various downstream applications. Typical yields of high copy plasmids are 2-6 mg from 400 mL of overnight culture in LB broth.
The GenElute Five-Minute Plasmid Miniprep Kit features an ultra-streamlined protocol resulting in high-quality plasmid DNA in about five minutes.

Recombinant E. coli overnight culture in LB (Luria-Bertani) broth is treated briefly (1-2 minutes) with a lysis reagent to effect rapid cell lysis and RNA degradation, without prior removal of culture medium. A binding solution is then mixed with the lysate, and DNA is captured on a silica-based binding column. Impurities are removed by a wash solution and bound plasmid DNA is then eluted in Tris buffer or water, ready for immediate use in various downstream applications. Typical yields of high copy plasmids are 2-6 μg from 400 μL of overnight culture in LB broth.

The GenElute Five-Minute Plasmid Miniprep Kits utilize a novel lysis procedure that eliminates the need to pellet cells from an overnight culture, remove growth media or clear a neutralized lysate. 400 μl of culture is added to a 2 mL collection tube and cells are lysed for 1−2 minutes. A Binding Solution is then added and the entire mixture is passed through a binding column. After washing the column to remove trace contaminants, the plasmid DNA is eluted in 10 mM Tris HCl, pH 8.5. Molecular biology water may be substituted as an elution reagent.

Application

GenElute Five-Minute Plasmid Miniprep Kit has been used in the isolation of plasmid.[1]
The GenElute Five-Minute Plasmid Miniprep Kit is intended for rapid preparation of plasmid DNA for use in capillary DNA sequencing, clone screening, restriction digestion, and PCR. For low copy plasmids, the kit can be used to prepare plasmid DNA for use in restriction digestion and PCR.

Features and Benefits

  • No pelleting cells or clearing lysates
  • Binding column works with any standard laboratory vacuum manifold
  • Up to 5 μg of plasmid DNA or enough for 15 sequencing reactions
  • DNA suitable for immediate capillary DNA sequencing

Other Notes

For additional information, please see www.sigma-aldrich.com/genelutehp.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1C - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

flash_point_f

69.8 °F

flash_point_c

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of TERT on the growth of fibrosarcoma via caspase-3, survivin and PKB
Qiuye Ma
Oncology Letters (2017)
Qiuye Ma et al.
Oncology letters, 14(2), 1939-1942 (2017-08-10)
The present study explored the effect of telomerase reverse transcriptase (TERT) on the growth and apoptosis of fibrosarcoma, and investigated the potential molecular signalling pathways underlying its effect. A plasmid was constructed in order to overexpress TERT and siRNA was
Effect of TERT on the growth of fibrosarcoma via caspase-3, survivin and PKB
Ma Q, et al.
Oncology Letters, 14(2), 1939-1942 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service