H9503
Butyl 4-hydroxybenzoate
Synonym(s):
p-Hydroxybenzoic acid n-butyl ester, Butyl paraben
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
194.23
Beilstein/REAXYS Number:
1103741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
bp
156-157 °C/3.5 mmHg (lit.)
mp
67-70 °C (lit.)
SMILES string
CCCCOC(=O)c1ccc(O)cc1
InChI
1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3
InChI key
QFOHBWFCKVYLES-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Guoyuan Wen et al.
The Journal of general virology, 96(Pt 6), 1219-1228 (2015-01-30)
Thermostable Newcastle disease virus (NDV) vaccines have been used widely to control Newcastle disease for village poultry flocks, due to their independence of cold chains for delivery and storage. To explore the potential use of thermostable NDV as a vaccine
Caroline Stremnitzer et al.
The Journal of investigative dermatology, 135(7), 1790-1800 (2015-02-24)
Papain is commonly used in food, pharmaceutical, textile, and cosmetic industries and is known to induce occupational allergic asthma. We have previously shown that the papain-like cysteine protease Dermatophagoides pteronyssinus 1 from house dust mite exhibits percutaneous sensitization potential. We
Jordan Shaw et al.
Reproductive toxicology (Elmsford, N.Y.), 28(1), 26-31 (2009-06-06)
Parabens, a class of preservatives routinely added to cosmetics, pharmaceuticals, and foods, have estrogenic properties. Given that intrauterine implantation of fertilized ova in inseminated females can be disrupted by minute levels of exogenous estrogens, we assessed the impact of parabens
A Beltran et al.
Analytica chimica acta, 677(1), 72-78 (2010-09-21)
In this paper we describe the synthesis, characterisation and use of two distinct molecularly imprinted polymers (MIPs) prepared using esters of p-hydroxybenzoic acid (parabens) as templates: one MIP was synthesised by precipitation polymerisation using a semi-covalent molecularly imprinting strategy with
Anna Viridén et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 77(1), 99-110 (2010-11-18)
This study investigates the effect of the chemical heterogeneity of hydroxypropyl methylcellulose (HPMC) on the release of model drug substances from hydrophilic matrix tablets. The hypothesis was that the release of drug substances could be influenced by possible interactions with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service