Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

FLMAS

Sigma-Aldrich

Mammalian Amino-terminal FLAG® Stable Expression Kit

Kit includes pFLAG-CMV-3 (E6783) and pFLAG-CMV-4 (E7158)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32161000

grade

for molecular biology

form

buffered aqueous solution

shipped in

wet ice

storage temp.

−20°C

General description

The Mammalian Amino-Terminal FLAG Stable Expression Kit provides the specialized FLAG® components needed for expression of a recombinant N-terminal FLAG fusion in mammalian cells. The pFLAG-CMV-3 and pFLAG-CMV-4 expression vectors encode the aminoglycoside phosphotransferase II gene (Neo) which confers resistance to aminoglycosides such as G418, allowing for selection of stable transfectants.

Vector Maps and Sequences

Application

The pFLAG-CMV-3 and pFLAG-CMV-4 Expression Vectors are suitable for transient or stable expression of secreted or cytoplasmic N-terminal FLAG® fusion proteins.

Components

  • pFLAG-CMV-3a Expression Vector (E8770) 20 μg
  • pFLAG-CMV-4a Expression Vector (E1775) 20 μg
  • pFLAG-CMV-3-BAPa Control Plasmid (C3972) 20 μg
  • pFLAG-CMV-4-BAPa Control Plasmid (C4722) 20 μg
  • C-CMV-24 Reverse Sequencing Primer (P5475) 1 μg
  • N-CMV-30 Forward Sequencing Primer (P5350) 1 μg
  • ANTI-FLAG M2 Monoclonal Antibody (F3165) 200 μg
  • ANTI-FLAG M5 Monoclonal Antibody (F4042) 200 μg
  • ANTI-FLAG M2 Affinity Gel - Freezer Safe (A2220) 1 ml
  • Met-FLAG-BAP Control Protein (P5975) 100 ug
  • FLAG Peptide (F3290) 4 mg

Principle

The FLAG® system is based on the FLAG marker octapeptide (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys). The FLAG sequence is hydrophilic and the last 5 amino acids (Asp-Asp-Asp-Asp-Lys) represent the target sequence of the protease enterokinase. FLAG fusion proteins are created by cloning the coding sequence of a protein of interest adjacent to the FLAG coding sequence in a FLAG expression vector, followed by expression in the appropriate host cell type.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service