Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E2632

Sigma-Aldrich

2-Ethoxyethanol

~99% (GC)

Synonym(s):

Ethyl glycol, Ethylene glycol monoethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein/REAXYS Number:
1098271
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

vapor density

3.1 (vs air)

vapor pressure

3.8 mmHg ( 20 °C)

assay

~99% (GC)

autoignition temp.

460 °F

expl. lim.

14 %

refractive index

n20/D 1.407 (lit.)

bp

135 °C (lit.)

mp

−90 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CCOCCO

InChI

1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3

InChI key

ZNQVEEAIQZEUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

104.0 °F - closed cup

flash_point_c

40 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J Traynor et al.
Toxicology and applied pharmacology, 218(2), 128-134 (2006-12-19)
Glycol ethers are solvents widely used alone and as mixtures in industrial and household products. Some glycol ethers have been shown to have a range of toxic effects in humans following absorption and metabolism to their aldehyde and acid metabolites.
David J Lockley et al.
Toxicology and applied pharmacology, 180(2), 74-82 (2002-04-24)
Percutaneous absorption and cutaneous metabolism of 2-ethoxyethanol were assessed in vivo and with an in vitro flow-through diffusion system. Topical application of undiluted (14)C-ethoxyethanol to occluded rat skin in vivo resulted in 25% of the dose being absorbed after 24
Chang Yong Yoon et al.
The Journal of veterinary medical science, 65(2), 207-212 (2003-03-26)
The effects of ethylene glycol monoethyl ether (EGEE) on testicular cell populations in rats were investigated by a flow cytometric method. Rats were administered by gavage with EGEE at the various doses of 0 (saline alone), 100, 200, 400, and
Gargi Bagchi et al.
International journal of andrology, 31(2), 269-274 (2008-01-09)
Methoxyacetic acid (MAA), the active biological oxidation product of the industrial solvent ethylene glycol monomethyl ether (EGME), causes acute toxicity in several species including humans. MAA primarily affects tissues with rapidly dividing cells and high rates of energy metabolism, including
Douglas J Fort et al.
Drug and chemical toxicology, 25(3), 293-308 (2002-08-14)
Currently, no standardized and well-validated alternative models exist for screening for progesterone-responsive endocrine disrupting chemicals (EDCs). Because of this, a rapid assay for evaluating progestin/antiprogestin activity using Xenopus oocyte germinal vesicle breakdown (GVBD) as a model was evaluated. Five compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service