Skip to Content
MilliporeSigma

14293

1-(4,6-Dichloro-1,3,5-triazin-2-yl)pyrene

BioReagent, suitable for fluorescence, ≥85% (HPCE)

Synonym(s):

1-(dichloro-1,3,5-triazinyl)pyrene, 4,6-Dichloro-2-(1-pyrenyl)-1,3,5-triazine, PyTC2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H9Cl2N3
CAS Number:
Molecular Weight:
350.20
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352116
EC Number:
221-754-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

assay

≥85% (HPCE)

form

powder

solubility

chloroform: soluble

fluorescence

λex 458 nm; λem 497 nm in DMF

suitability

suitable for fluorescence

SMILES string

Clc1nc(Cl)nc(n1)-c2ccc3ccc4cccc5ccc2c3c45

InChI

1S/C19H9Cl2N3/c20-18-22-17(23-19(21)24-18)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9H

InChI key

ZDMAOBNCIJTNCB-UHFFFAOYSA-N

Application

1-(4,6-Dichloro-1,3,5-triazin-2-yl)pyrene is a fluorescent probe with high quantum efficiency, exhibiting single exponential decay kinetics. The radiative fluorescence lifetime is almost constant and independent on molecular surroundings allowing this probe to be utilized in a range of microsurroundings with exciting radiation at a constant wavelength (in the blue spectral range). Its fluorescence emission demonstrates a large solvatochromic shift which changes with solvent polarity. For example, the maximum shifts from 435 nm in low polarity n-heptane to 492 nm in high polarity acetonitrile. This product is only sparingly soluble in water .


pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library